missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-3’,5’-cyclic Monophosphate, TRC

Product Code. 30325698
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325698 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325698 Supplier Toronto Research Chemicals Supplier No. TRCA2804555G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adenosine-3',5'-cyclic Monophosphate
CAS 60-92-4
Molecular Formula C10 H12 N5 O6 P
Quantity 5 g
Synonym Adenosine 3',5'-Cyclic Phosphate, 3',5'-AMP, 45: PN: US20030109453 SEQID: 44 Claimed sequence, AMPc, Adenosine 3',5'-Cyclophosphate, Adenosine 3',5'-Monophosphate, Adenosine 3',5'-Phosphate, Adenosine Cyclic 3',5'-Monophosphate, Adenosine Cyclic Monophosphate, Cyclic 3',5'-AMP, Cyclic 3',5'-Adenylic Acid, Cyclic AMP, Cyclic Adenosine 3',5'-Monophosphate, Cyclic Adenosine 3',5'-Phosphate, NSC 143670, NSC 94017, cAMP
InChI Formula InChI=1S/C10H12N5O6P/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(16)7-4(20-10)1-19-22(17,18)21-7/h2-4,6-7,10,16H,1H2,(H,17,18)(H2,11,12,13)/t4-,6-,7-,10-/m1/s1
SMILES Nc1ncnc2c1ncn2[C@@H]3O[C@@H]4COP(=O)(O)O[C@H]4[C@H]3O
IUPAC Name (4aR,6R,7R,7aS)-6-(6-aminopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
Molecular Weight (g/mol) 329.21
Formula Weight 329.0525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.