Learn More
Thermo Scientific Chemicals Adenosine 3',5'-cyclic monophosphate, 99+%
Tienda Thermo Scientific Chemicals ProductosDescripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Absorbance | lambda max. 260nm,pH 7,0.77 to 0.83 (A250/A260),0.13 to 0.19 (A280/A260) |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | 260.0°C |
| Assay Percent Range | 99% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Beilstein | 26, IV, 3618 |
| Merck Index | 15, 2701 |
| Specific Rotation | − 53.70 |
| Mostrar más |
Seguridad y manipulación
- Adenosine 3'-,5'-cyclic monophosphate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.