missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine 3',5'-cyclic monophosphate, 99+%

Product Code. 10144900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144900 500 mg Glass bottle 500mg
10432611 1 g Glass bottle 1g
10781915 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10144900 Supplier Thermo Scientific Chemicals Supplier No. 225805000
  • ONLINE EXCLUSIVE PRICE
    £125.00
    £112.65 / 500mg
    Save £12.35
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 60-92-4 | C10H12N5O6P | 329.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-92-4
Molecular Formula C10H12N5O6P
Molecular Weight (g/mol) 329.21
MDL Number MFCD00005845
InChI Key IVOMOUWHDPKRLL-YPLCUDRINA-N
Synonym camp, cyclic amp, adenosine 3',5'-cyclic monophosphate, 3',5'-cyclic amp, adenosine 3',5'-phosphate, adenosine 3',5'-cyclophosphate, cyclic 3',5'-amp, adenosine cyclic monophosphate, adenosine-3',5'-cyclophosphate, cyclic adenosine monophosphate
PubChem CID 6076
ChEBI CHEBI:17489
SMILES NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

Absorbance lambda max. 260nm,pH 7,0.77 to 0.83 (A250/A260),0.13 to 0.19 (A280/A260)
CAS Min % 99.0
CAS Max % 100.0
Melting Point 260.0°C
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 26, IV, 3618
Merck Index 15, 2701
Specific Rotation − 53.70
Solubility Information Solubility in water: 50mg/mL
IUPAC Name (4aR,6R,7R,7aS)-6-(6-aminopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
Formula Weight 329.2
Percent Purity 99+%
Chemical Name or Material Adenosine 3′,5′-cyclic monophosphate
Show More Show Less
compliance-icons
Product Identifier
  • Adenosine 3'-,5'-cyclic monophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.