missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-2',3'-cyclic monophosphate sodium salt, 97%

Product Code. 15470137
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470137 100 mg 100mg
15480137 250 mg 250mg
15490137 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470137 Supplier Thermo Scientific Alfa Aesar Supplier No. J60558.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37063-35-7 | C10H11N5NaO6P | 351.19 g/mol

Adenosine-2’,3’-cyclic Monophosphate is an Adenosine (A280400) derivative useful as anticancer agent. It is also used as a Pharmaceutical intermediate, pharmaceutical research and chemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-2’,3’-cyclic Monophosphate is an Adenosine (A280400) derivative useful as anticancer agent. It is also used as a Pharmaceutical intermediate, pharmaceutical research and chemical analysis.

Solubility
It is soluble in DMSO, water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37063-35-7
Molecular Formula C10H11N5NaO6P
Molecular Weight (g/mol) 351.19
MDL Number MFCD00005757
InChI Key VSDSIACSNXHGOV-GWKNMROSNA-M
Synonym adenosine-2 inverted exclamation mark ,3 inverted exclamation mark-cyclic monophosphate sodium salt
PubChem CID 131698406
IUPAC Name [(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;sodium
SMILES [Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1

Specifications

Melting Point 241°C to 243°C
Color White
Quantity 100 mg
Solubility Information It is soluble in DMSO,water.
Formula Weight 351.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Adenosine-2',3'-cyclic monophosphate sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.