missing translation for 'onlineSavingsMsg'
Learn More

Novacyt - Microgen Bioproducts Adenoscreen Latex

Product Code. 17353211 Shop All Novacyt Group Products
Change view
Click to view available options
Clia Complexity:
Non-waived
Unit Size:
25 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Clia Complexity unitSize
17353211 Non-waived 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17353211 Supplier Novacyt - Microgen Bioproducts Supplier No. M81CE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Microgen Bioproducts Adenoscreen Latex - Rapid and simple latex agglutination test for the detection of Adenovirus antigen in faecal specimens.

  • A simple and rapid latex agglutination test for the detection of Rotavirus antigen in facal specimens.

Specifications

Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
Control Sets M81c Positive control - 1.0 mL
CE Marker Yes
Clia Complexity Non-waived
Detectable Analytes Adenovirus Antibodies (Rabbit)
Format 4 Bottles (2.5 mL, 2.5 mL, 1.5 mL, 50 mL)
Sample Type Suspension of faecal specimen
Sensitivity 0.96
Type Latex Agglutination Test
For Use With (Application) The Detection of Adenovirus Antigen in Faecal Specimens
For Use With (Equipment) Laboratory Centrifuge, Stoppered or Screw-Capped Tubes
Includes 50 tests
Material Polyethylene Dropper Bottles
No. of Reactions 2 per test
Organism Group Animal
Packaging Type Carton
Quantity 4 Bottles (2.5 mL, 2.5 mL, 1.5 L, 50 mL)
Reagent Type M81a - Test reagent - 2.5 mL - Contains (rabbit) Adenovirus antibody-sensitised latex particle
Species Adenovirus
Specificity 0.99
Target Organism Class Eukaryota
Test Time 12 minutes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.