missing translation for 'onlineSavingsMsg'
Learn More

Adenine, TRC

Product Code. 30137916
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137916 25 g 25g
18010530 1 g 1g
18042530 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137916 Supplier Toronto Research Chemicals Supplier No. TRCA28048025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5 H5 N5
Molecular Weight (g/mol) 135.13
Synonym 9H-Purin-6-amine, 1H-Purin-6-amine (9CI), Adenine (8CI), (9H-Purin-6-yl)amine, 1,6-Dihydro-6-iminopurine, 3,6-Dihydro-6-iminopurine, 3H-Purin-6-amine, 6-Amino-1H-purine, 6-Amino-3H-purine, 6-Amino-9H-purine, 6-Aminopurine, 7H-Imidazo[4,5-c]pyridazin-3-amine, 7H-Purin-6-amine, 9H-Adenine, Adeninimine, Leuco 4, NSC 14666, Pedatisectine B, Vitamin B4
IUPAC Name 7H-purin-6-amine
SMILES Nc1ncnc2nc[nH]c12

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)
Formula Weight 135.0545
Percent Purity >95%
Chemical Name or Material Adenine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.