missing translation for 'onlineSavingsMsg'
Learn More

Adenine Nucleotide Translocator 2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-92630-0.02ml

 View more versions of this product

Product Code. 18692292

  • £151.00 / 0.02mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Adenine Nucleotide Translocator 2 Polyclonal antibody specifically detects Adenine Nucleotide Translocator 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adenine Nucleotide Translocator 2
Polyclonal
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
Adenine nucleotide translocator 2, adenine nucleotide translocator 2 (fibroblast), ADP, ADP/ATP carrier protein, ADP/ATP translocase 2, ANT2AAC2, 2F1, ATP carrier protein 2, ATP carrier protein, fibroblast isoform, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 5, Solute carrier family 25 member 5, T2, T3ANT 2
A synthetic peptide corresponding to a sequence within amino acids 151-250 of human Adenine Nucleotide Translocator 2 (NP_001143.2). KGMLPDPKNTHI
0.02 mL
Core ESC Like Genes, Stem Cell Markers
292
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.