missing translation for 'onlineSavingsMsg'
Learn More

Adenine Nucleotide Translocator 2 Antibody (1D7), Novus Biologicals™

Product Code. 18354518 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354518 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354518 Supplier Novus Biologicals Supplier No. H00000292M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenine Nucleotide Translocator 2 Monoclonal antibody specifically detects Adenine Nucleotide Translocator 2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenine Nucleotide Translocator 2
Applications ELISA
Classification Monoclonal
Clone 1D7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH56160
Gene Alias Adenine nucleotide translocator 2, adenine nucleotide translocator 2 (fibroblast), ADP, ADP/ATP carrier protein, ADP/ATP translocase 2, ANT2AAC2, 2F1, ATP carrier protein 2, ATP carrier protein, fibroblast isoform, solute carrier family 25 (mitochondrial carrier, adenine nucleotidetranslocator), member 5, Solute carrier family 25 member 5, T2, T3ANT 2
Host Species Mouse
Immunogen SLC25A5 (AAH56160, 1 a.a. ~ 298 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTDAAVSFAKDFLAGGVAAAISKTAVAPIERVKLLLQVQHASKQITADKQYKGIIDCVVRIPKEQGVLSFWRGNLANVIRYFPTQALNFAFKDKYKQIFLGGVDKRTQFWRYFAGNLASGGAAGATSLCFVYPLDFARTRLAADVGKAGAEREFRGLGDCLVKIYKSDGIKGLYQGFNVSVQGIIIYRAAYFGIYDTAKGMLPDPKNTHIVISWMIAQTVTAVAGLTSYPFDTVRRRMMMQSGRKGTDIMYTGTLDCWRKIARDEGGKAFFKGAWSNVLRGMGGAFVLVLYDEIKKYT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 292
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.