missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 6055-72-7 |
| Color | White |
| Molecular Formula | C5H8ClN5O |
| MDL Number | MFCD11046306 |
| Quantity | 5 g |
| Synonym | 7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride |
| InChI Key | MYRDTAUFFBYTHA-UHFFFAOYSA-N |
| SMILES | O.Cl.NC1=C2NC=NC2=NC=N1 |
| IUPAC Name | 7H-purin-6-amine hydrate hydrochloride |
| Molecular Weight (g/mol) | 189.60 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?