missing translation for 'onlineSavingsMsg'
Learn More

Adenine Hemisulfate, TRC

Product Code. 30313847
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313847 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313847 Supplier Toronto Research Chemicals Supplier No. TRCA28036525G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adenine Sulphate
CAS 321-30-2
Molecular Formula 2[C5H5N5].SO4H2
Quantity 25 g
Synonym 7H-Purin-6-amine Sulphate, Adenine Sulphate, 9H-Purin-6-amine, sulfate (2:1), 1H-Purin-6-amine, sulfate (2:1) (9CI), Adenine, sulfate (2:1) (8CI), Adenine hemisulfate, Adenine sulfate, Adeninium sulfate
InChI Formula InChI=1S/2C5H5N5.H2O4S/c2*6-4-3-5(9-1-7-3)10-2-8-4;1-5(2,3)4/h2*1-2H,(H3,6,7,8,9,10);(H2,1,2,3,4)
SMILES Nc1ncnc2[nH]cnc12.Nc3ncnc4[nH]cnc34.OS(=O)(=O)O
IUPAC Name 9H-purin-6-amine;sulfuric acid
Molecular Weight (g/mol) 368.33
Formula Weight 368.0764
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.