missing translation for 'onlineSavingsMsg'
Learn More

Melford Adenine [6-Aminopurine] [Vitamin B4] USP

Product Code. p-200087117
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 30017950

Brand: Melford A11500100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A nucleobase with a variety of roles in biochemistry including cellular respiration, in the form of both the energy-rich adenosine triphosphate and the cofactors nicotinamide adenine dicucleotide and flavin adenine dinucleotide, and protein synthesis, as a chemical component of DNA and RNA.

A nucleobase with a variety of roles in biochemistry including cellular respiration, in the form of both the energy-rich adenosine triphosphate and the cofactors nicotinamide adenine dicucleotide and flavin adenine dinucleotide, and protein synthesis, as a chemical component of DNA and RNA.

Specifications

CAS 73-24-5
Molecular Formula C5H5N5
Quantity 100 g
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.