missing translation for 'onlineSavingsMsg'
Learn More

Melford Adenine [6-Aminopurine] [Vitamin B4] USP

Product Code. 30017952
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017952 25 g 25g
30017950 100 g 100g
30017951 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30017952 Supplier Melford Supplier No. A1150025.0
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A nucleobase with a variety of roles in biochemistry including cellular respiration, in the form of both the energy-rich adenosine triphosphate and the cofactors nicotinamide adenine dicucleotide and flavin adenine dinucleotide, and protein synthesis, as a chemical component of DNA and RNA.

A nucleobase with a variety of roles in biochemistry including cellular respiration, in the form of both the energy-rich adenosine triphosphate and the cofactors nicotinamide adenine dicucleotide and flavin adenine dinucleotide, and protein synthesis, as a chemical component of DNA and RNA.

Specifications

CAS 73-24-5
Molecular Formula C5H5N5
Quantity 25 g
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.