missing translation for 'onlineSavingsMsg'
Learn More

Ademethionine 1,4-Butanedisulfonate, TRC

Product Code. 17998024
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17998024 10 mg 10mg
17969564 500 mg 500mg
17938914 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17998024 Supplier Toronto Research Chemicals Supplier No. TRCA28035510MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101020-79-5
Molecular Formula 2 C15 H22 N6 O5 S . 3 C4 H10 O6 S2
Synonym Adenosine, 5'-[[(3S)-3-amino-3-carboxypropyl]methylsulfonio]-5'-deoxy-, 1,4-butanedisulfonate (salt), 1,4-butanedisulfonate (salt) (2:1:2)∣Adenosine, 5'-[(3-amino-3-carboxypropyl)methylsulfonio]-5'-deoxy-, (3S)-, 1,4-butanedisulfonate (salt), 1,4-butanedisulfonate (salt) (2:1:2)∣1,4-Butanedisulfonic acid, compd. with (3S)-5'-[(3-amino-3-carboxypropyl)methylsulfonio]-5'-deoxyadenosine 1,4-butanedisulfonate (salt) (2:2:1)∣1,4-Butanedisulfonic acid, compd. with 5'-[[(3S)-3-amino-3-carboxypropyl]methylsulfonio]-5'-deoxyadenosine 1,4-butanedisulfonate (salt) (2:2:1) (9CI)∣1,4-Butanedisulfonic acid, ion(2-), bis[(3S)-5'-[(3-amino-3-carboxypropyl)methylsulfonio]-5'-deoxyadenosine], 1,4-butanedisulfonate (1:2) (salt)∣1,4-Butanedisulfonic acid, ion(2-), bis[5'-[[(3S)-3-amino-3-carboxypropyl]methylsulfonio]-5'-deoxyadenosine], 1,4-butanedisulfonate (1:2) (salt) (9CI)∣Ademethionine 1,4-Butanedisulfonate

Specifications

Format Neat
Molecular Weight (g/mol) 1451.62
Shipping Condition Room Temperature
Purity >95% (HPLC)
Quantity 10 mg
Chemical Name or Material Ademethionine 1,4-Butanedisulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.