missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adefovir dipivoxil, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Adefovir dipivoxil, an adenosine analogue, is an oral prodrug of the nucleoside reverse transcriptase inhibitor Adefovir. Adefovir dipivoxil inhibits both the wild type and HBV Lamivudine-resistant strains. Adefovir dipivoxil shows anti-orthopoxvirus activity.
Specifications
Specifications
| Chemical Name or Material | Adefovir dipivoxil |
| CAS | 142340-99-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H32N5O8P |
| Quantity | 50 mg |
| Synonym | GS 0840 |
| Solubility Information | DMSO : ≥ 100 mg/mL (199.41 mM) ∣H2O : 0.67 mg/mL (1.34 mM; Need ultrasonic) |
| SMILES | NC1=C2N=CN(CCOCP(OCOC(C(C)(C)C)=O)(OCOC(C(C)(C)C)=O)=O)C2=NC=N1 |
| Molecular Weight (g/mol) | 501.47 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction