missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adefovir Dipivoxil Dimer, TRC
Specifications
Specifications
| Chemical Name or Material | Adefovir Dipivoxil Dimer |
| CAS | 323201-05-4 |
| Molecular Formula | C41 H64 N10 O16 P2 |
| Quantity | 2.5 mg |
| Synonym | Adefovir Dipivoxil Impurity B, 2,2-Dimethyl-propanoic Acid Methylenebis[imino-9H-purine-6,9-diyl-2,1-ethanediyloxymethylenephosphinylidynebis(oxymethylene)] Ester |
| InChI Formula | InChI=1S/C41H64N10O16P2/c1-38(2,3)34(52)60-22-64-68(56,65-23-61-35(53)39(4,5)6)26-58-15-13-50-20-48-28-30(44-18-46-32(28)50)42-17-43-31-29-33(47-19-45-31)51(21-49-29)14-16-59-27-69(57,66-24-62-36(54)40(7,8)9)67-25-63-37(55)41(10,11)12/h18-21H,13-17,22-27H2,1-12H3,(H,42,44,46)(H,43,45,47) |
| SMILES | CC(C)(C)C(=O)OCOP(=O)(COCCn1cnc2c(NCNc3ncnc4c3ncn4CCOCP(=O)(OCOC(=O)C(C)(C)C)OCOC(=O)C(C)(C)C)ncnc12)OCOC(=O)C(C)(C)C |
| IUPAC Name | [2-[6-[[[9-[2-[bis(2,2-dimethylpropanoyloxymethoxy)phosphorylmethoxy]ethyl]purin-6-yl]amino]methylamino]purin-9-yl]ethoxymethyl-(2,2-dimethylpropanoyloxymethoxy)phosphoryl]oxymethyl 2,2-dimethylpropanoate |
| Molecular Weight (g/mol) | 1014.952 |
| Formula Weight | 1014.398 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?