missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adecatumumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559368
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Adecatumumab is a recombinant human monoclonal antibody of the IgG1 subclass with a binding specificity to epithelial cell adhesion molecule (EpCAM). EpCAM (CD326) is a cell surface protein that is frequently expressed at high level on most solid tumor types, including prostate, breast, colon, gastric, ovarian, pancreatic and lung cancer. Adecatumumab (MT201) is a recombinant human monoclonal antibody of the IgG1 subclass with a binding specificity to epithelial cell adhesion molecule (EpCAM). EpCAM (CD326) is a cell surface protein that is frequently expressed at high level on most solid tumor types, including prostate, breast, colon, gastric, ovarian, pancreatic and lung cancer. Overexpression of EpCAM has been shown to promote the proliferation, migration and invasiveness of breast cancer cells. Moreover, expression of EpCAM is associated with decreased survival in a number of cancer indications, including breast, gall bladder, bile duct, ovarian and ampullary pancreatic cancer. EpCAM has recently been shown to be expressed on cancer stem cells from breast, prostate, colon and pancreatic cancer. Clinical studies have shown a favorable safety profile and that clinical activity depends on the EpCAM target expression.
Specifications
| Adecatumumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MT201 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction