missing translation for 'onlineSavingsMsg'
Learn More

ADE2 Antibody (4F4), Novus Biologicals™

Product Code. 18326028 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326028 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326028 Supplier Novus Biologicals Supplier No. H00010606M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADE2 Monoclonal antibody specifically detects ADE2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADE2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4F4
Conjugate Unconjugated
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH10273
Gene Alias ADE2, AIR carboxylase, AIRCADE2H1, DKFZp781N1372, MGC1343, MGC5024, multifunctional protein ADE2, PAIS, phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazolesuccinocarboxamide synthetase, SAICAR synthetase
Host Species Mouse
Immunogen PAICS (AAH10273, 1 a.a. ~ 425 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MATAEVLNIGKKLYEGKTKEVYELLDSPGKVLLQSKDQITAGNAARKNHLEGKAAISNKITSCIFQLLQEAGIKTAFTRKCGETAFIAPQCEMIPIEWVCRRIATGSFLKRNPGVKEGYKFYPPKVELFFKDDANNDPQWSEEQLIAAKFCFAGLLIGQTEVDIMSHATQAIFEILEKSWLPQNCTLVDMKIEFGVDVTTKEIVLADVIDNDSWRLWPSGDRSQQKDKQSYRDLKEVTPEGLQMVKKNFEWVAERVELLLKSESQCRVVVLMGSTSDLGHCEKIKKACGNFGIPCELRVTSAHKGPDETLRIKAEYEGDGIPTVFVAVAGRSNGLGPVMSGNTAYPVISCPPLTPDWGVQDVWSSLRLPSGLGCSTVLSPEGSAQFAAQIFGLSNHLVWSKLRASILNTWISLKQADKKIRECNL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10606
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.