Learn More
Memmert™ Additional Stainless Steel Grid, electro-polished, For Ich750
Product Code. 16386747 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
Models 450 and 750 Ovens
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
16386747 Models 450 and 750 Ovens Each
1 options
Product Code. 16386747 Supplier Memmert™ Supplier No. ICH750E20182
Ideal for ramp operation with rapid temperature changes during incubation, breeding or storage.
  • For use with compressor-cooled incubator models 450 and 750
  • Grid pattern allows for airflow
  • Manufactured from electro-polished stainless steel
Specifications
For Use With (Equipment) Models 450 and 750 Ovens
Material Stainless Steel
Type Grid