Learn More
Memmert™ Additional Reinforced Stainless Steel Grid (For Reinforced Chamber Option), For Uf450Ts
Product Code. 16394157 Shop All Memmert Products
Click to view available options
Type:
Reinforced Grid
Unit Size:
Each
Product Code. 16394157
Brand: Memmert™ UF450TSB32190
Only null left
Add to basket
Add to basket
Includes
Guide Bars And Fixing Screws
Use these grids with Memmert ovens. Memmert™ Reinforced Grids are designed to hold up to 60 kg.
  • Reinforced stainless steel grid
  • Max. loading 60 kg
  • From size 450 with guide bars and fixing screws and only in connection with option K1
Specifications
Type Reinforced Grid
Weight (Metric) 60 kg