Learn More
Memmert™ Additional Reinforced Stainless Steel Grid (For Reinforced Chamber Option), For Ich260
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
Model 260
Unit Size:
Each
Description
- Reinforced stainless steel grid
- Max. loading 60 kg
- From size 450 with guide bars and fixing screws and only in connection with option K1
Specifications
Specifications
| Capacity (Metric) Shelf | 60 kg |
| For Use With (Equipment) | Model 260 |
| Material | Stainless Steel |
| Type | Reinforced Grid |
| Model | 260 |