missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Additional Perforated Stainless Steel Shelf for UN75plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN75plus Oven
Unit Size:
Each
Description
- Perforated Stainless Steel Shelf
- Accessory for Memmert ovens and incubators
Specifications
Specifications
| For Use With (Equipment) | UN75plus Oven |
| Type | Perforated Shelf |
| Material | Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction