missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Additional Perforated Stainless Steel Shelf for UN450

Product Code. 17590156 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN450 Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17590156 UN450 Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17590156 Supplier Memmert™ Supplier No. UN450B00328
  • ONLINE EXCLUSIVE PRICE
    £213.00
    £208.14 / Pack of 1
    Save £4.86
    2% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Shelves with a perforated stainless steel construction for ovens and incubators.

  • Perforated Stainless Steel Shelf
  • Accessory for Memmert ovens and incubators

Specifications

For Use With (Equipment) UN450 Oven
Type Perforated Shelf
Material Stainless Steel
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.