missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADCY3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19492-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.Specifications
| ADCY3 | |
| Polyclonal | |
| Unconjugated | |
| ADCY3 | |
| AC III, AC3, ADCY3, adenylate cyclase 3, Adenylate cyclase type 3, Adenylate cyclase type III, Adenylyl cyclase 3, ATP pyrophosphate lyase 3, KIAA0511 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104111, 109, 64508 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60266, P21932, Q8VHH7 | |
| Adcy3 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction