Learn More
ADCY3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19492-1-AP-150UL
 View more versions of this product
Product Code. 16829014
  • £349.00 / 150µL
Description
This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.
Specifications
Specifications
ADCY3
Polyclonal
Unconjugated
ADCY3
AC III, AC3, ADCY3, adenylate cyclase 3, Adenylate cyclase type 3, Adenylate cyclase type III, Adenylyl cyclase 3, ATP pyrophosphate lyase 3, KIAA0511
Rabbit
Antigen Affinity Chromatography
RUO
104111, 109, 64508
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.65 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60266, P21932, Q8VHH7
Adcy3
Peptide
150 μL
Primary
Human, Mouse, Rat