missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADCY2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527396
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, IMR32, HCT116. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ADCY2 is a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate(cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex.
Specifications
| ADCY2 | |
| Polyclonal | |
| Unconjugated | |
| ADCY2 | |
| 3',5'-cyclic AMP synthetase; AC2; Adcy2; adenylate cyclase 2; adenylate cyclase 2 (brain); adenylate cyclase II; adenylate cyclase type 2; Adenylate cyclase type II; adenylyl cyclase 2; ATP pyrophosphate-lyase 2; HBAC2; KIAA1060; mKIAA1060; type II adenylate cyclase; type II adenylyl cyclase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q08462 | |
| ADCY2 | |
| Recombinant protein encompassing a sequence within the center region of human Adenylate cyclase 2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction