missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADCY1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£139.05 - £349.00
Specifications
| Antigen | ADCY1 |
|---|---|
| Concentration | 0.46 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16816515
|
Proteintech
55067-1-AP-20UL |
20 μL |
£139.05
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16806515
|
Proteintech
55067-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to ADCY1.
ADCY1 belongs to the adenylyl cyclase class-4/guanylyl cyclase family. It is a calmodulin-sensitive adenylyl cyclase. ADCY1 may be involved in regulatory processes in the central nervous system. It may play a role in memory acquisition and learning.Specifications
| ADCY1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q08828 | |
| 107 | |
| Peptide | |
| Primary | |
| -20°C | |
| ADCY1 |
| 0.46 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AC1, ADCY1, adenylate cyclase 1 (brain), Adenylate cyclase type 1, Adenylate cyclase type I, Adenylyl cyclase 1, ATP pyrophosphate lyase 1 | |
| ADCY1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title