missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADCK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17579-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
ADCK5 is member of the AarF domain containing kinase family.Specifications
ADCK5 | |
Polyclonal | |
Unconjugated | |
ADCK5 | |
ADCK5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
203054, 268822, 362943 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3MIX3, Q80V03 | |
Adck5 | |
ADCK5 Fusion Protein Ag11094 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |