missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADCK5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555943
This item is not returnable.
View return policy
Description
Immunogen sequence: VLLDHGLYQF LEEKDRAALC QLWRAIILRD DAAMRAHAAA LGVQDYLLFA EMLMQRPVRL GQLWGSHLLS REEAAYMVDM ARERFEAVMA VLRELP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.
ADCK5 is member of the AarF domain containing kinase family.
Specifications
| ADCK5 | |
| Polyclonal | |
| Unconjugated | |
| ADCK5 | |
| A230108P17; aarF domain containing kinase 5; Adck5; Uncharacterized aarF domain-containing protein kinase 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 203054 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3MIX3 | |
| ADCK5 | |
| Recombinant protein corresponding to Human ADCK5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction