missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADCK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522180
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (90%), Rat (89%), Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein with two copies of a domain found in protein kinases. The encoded protein has a complete protein kinase catalytic domain, and a truncated domain that contains only the active and binding sites of the protein kinase domain, however, it is not known whether the protein has any kinase activity. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| ADCK4 | |
| Polyclonal | |
| Unconjugated | |
| COQ8B | |
| 0610012P18Rik; aarF domain containing kinase 4; aarF domain-containing protein kinase 4; ADCK4; Atypical kinase COQ8B, mitochondrial; Coenzyme Q protein 8B; coenzyme Q8B; COQ8B; FLJ12229; NPHS9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79934 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q96D53 | |
| COQ8B | |
| Recombinant protein encompassing a sequence within the center region of human ADCK4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction