missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ADCK3, GST Tag Recombinant Protein

Product Code. 17178785
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 17178785

Brand: Invitrogen™ A33372

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This ADCK3 recombinant human protein (156-end) was expressed in insect cells. The purified catalytic domain (135-end) of ADCK3 can be used in a number of different applications, but has been specifically validated for use with our LanthaScreen™ Kinase binding assay technology to identify novel kinase inhibitors. ADCK3 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Catalytic.

ADCK3 serine/threonine kinase is a mitochondrial protein similar to yeast ABC1 that is a member of the ADCK family. CABC1 encodes a mitochondrial protein similar to yeast ABC1, which functions in an electron-transferring membrane protein complex in the respiratory chain. It is not related to the family of ABC transporter proteins. Expression of this gene is induced by the tumor suppressor p53 and in response to DNA damage, and inhibiting its expression partially suppresses p53-induced apoptosis. Alternatively spliced transcript variants have been found.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NI60
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 10mM glutathione, 0.1mM PMSF, 0.25mM DTT, 0.1mM EDTA, 50mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 56997
Molecular Weight (g/mol) 82.3 kDa
Name Human ADCK3, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 4632432J16Rik; aarF domain containing kinase 3; aarF domain-containing protein kinase 3; Adck3; AI462003; ARCA2; atypical kinase ADCK3, mitochondrial; Atypical kinase COQ8A, mitochondrial; Cabc1; Chaperone activity of bc1 complex-like; chaperone activity of bc1 complex-like, mitochondrial; chaperone, ABC1 activity of bc1 complex homolog; chaperone, ABC1 activity of bc1 complex like; Chaperone-ABC1-like; Coenzyme Q protein 8A; coenzyme Q8 homolog; coenzyme Q8A; COQ10D4; COQ8; COQ8A; mKIAA0451; PP265; SCAR9
Common Name CABC1
Gene Symbol COQ8A
KinaseFamily ACK Kinase Family
Protein Length 135-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 135-end
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Recombinant, Catalytic
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.