Learn More
Invitrogen™ Human ADCK3, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This ADCK3 recombinant human protein (156-end) was expressed in insect cells. The purified catalytic domain (135-end) of ADCK3 can be used in a number of different applications, but has been specifically validated for use with our LanthaScreen™ Kinase binding assay technology to identify novel kinase inhibitors. ADCK3 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Catalytic.
Specifications
Specifications
| Accession Number | Q8NI60 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 25% glycerol, 10mM glutathione, 0.1mM PMSF, 0.25mM DTT, 0.1mM EDTA, 50mM NaCl and no preservative; pH 7.5 |
| Gene ID (Entrez) | 56997 |
| Molecular Weight (g/mol) | 82.3 kDa |
| Name | Human ADCK3, GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.