missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADCK2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15672995
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15672995 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15672995 Supplier Invitrogen™ Supplier No. PA548701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADCK2 Polyclonal Antibody for Western Blot

The function of this protein is not yet clear. It is not known if it has protein kinase activity and what type of substrate it would phosphorylate (Ser, Thr or Tyr).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADCK2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADCK2
Gene Accession No. Q6NSR3
Gene Alias AA408112; AARF; aarF domain containing kinase 2; Adck2; BB006057; MGC20727; putative ubiquinone biosynthesis protein AarF; uncharacterized aarF domain-containing protein kinase 2
Gene Symbols ADCK2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 232-259 amino acids from the Central region of mouse Adck2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57869
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.