missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADC Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67884-1-IG-20UL
This item is not returnable.
View return policy
Description
The ADC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADC. This antibody recognizes human, mouse, pig, rat antigen. The ADC antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| ADC | |
| Monoclonal | |
| 1000 μg/mL | |
| AZI2; KIAA1945; ODC-p; ODC1L | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| 48-50 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1A2B11 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 113451 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction