missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114193
This item is not returnable.
View return policy
Description
Immunogen sequence: RPFPACSFAP AAAPQAVRAG AVRKLDADED GLPYLCTGYD LYVTREPCAM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
This gene encodes a subunit of a tRNA-specific adenosine deaminase. This heterodimeric enzyme converts adenosine to inosine in the tRNA anticodon. A mutation in this gene causes a syndrome characterized by intellectual disability and strabismus. This gene shares its 5' exon with the overlapping gene, secretory carrier membrane protein 4. ADAT3 (Adenosine Deaminase TRNA Specific 3) is a Protein Coding gene. Diseases associated with ADAT3 include Neurodevelopmental Disorder With Brain Abnormalities, Poor Growth, And Dysmorphic Facies and Cone-Rod Dystrophy 10. Among its related pathways are tRNA processing and Processing of Capped Intron-Containing Pre-mRNA. Gene Ontology (GO) annotations related to this gene include hydrolase activity. An important paralog of this gene is ADAT2.
Specifications
| ADAT3 | |
| Polyclonal | |
| Unconjugated | |
| ADAT3 | |
| ADAT3; adenosine deaminase, tRNA specific 3; adenosine deaminase, tRNA-specific 3; adenosine deaminase, tRNA-specific 3, TAD3 homolog; FWP005; MRT36; MST121; MSTP121; Probable inactive tRNA-specific adenosine deaminase-like protein 3; S863-5; TAD3; tRNA-specific adenosine deaminase 3 homolog; tRNA-specific adenosine deaminase-like protein 3; tRNA-specific adenosine-34 deaminase subunit ADAT3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113179 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96EY9 | |
| ADAT3 | |
| Immunogen sequence: RPFPACSFAP AAAPQAVRAG AVRKLDADED GLPYLCTGYD LYVTREPCAM (aa 244-293). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction