missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAT3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99169-50ul
This item is not returnable.
View return policy
Description
ADAT3 Polyclonal antibody specifically detects ADAT3 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| ADAT3 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| adenosine deaminase, tRNA-specific 3, TAD3 homolog (S. cerevisiae), FWP005, MST121, MSTP121, S863-5, TAD3, tRNA-specific adenosine deaminase 3 homolog, tRNA-specific adenosine deaminase-like protein 3, tRNA-specific adenosine-34 deaminase subunit ADAT3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ADAT3. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 113179 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction