missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13621-1-AP-20UL
This item is not returnable.
View return policy
Description
Probably participates in deamination of adenosine-34 to inosine in many tRNAs.Specifications
| ADAT2 | |
| Polyclonal | |
| Unconjugated | |
| ADAT2 | |
| ADAT2, DEADC1, dJ20N2, TAD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 134637, 361453, 66757 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P6J0, Q7Z6V5 | |
| Adat2 | |
| ADAT2 Fusion Protein Ag4529 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction