Learn More
ADAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541835
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: RLVPCGAAIS WSAVPEQPLD VTANGFPQGT TKKTIGSLQA RSQISKVELF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
ADAT1 Polyclonal antibody specifically detects ADAT1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ADAT1 | |
Polyclonal | |
Unconjugated | |
ADAT1 | |
Adat1; Adenosine deaminase; adenosine deaminase acting on tRNA; adenosine deaminase tRNA specific 1; adenosine deaminase, tRNA specific 1; adenosine deaminase, tRNA-specific 1; HADAT1; mADAT1; MMADAT1; tRNA-specific 1; tRNA-specific adenosine deaminase 1; tRNA-specific adenosine-37 deaminase; zgc:162299 | |
Rabbit | |
Protein A | |
RUO | |
23536 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BUB4 | |
ADAT1 | |
Synthetic peptide directed towards the C-terminal of human ADAT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |