Learn More
ADAR1 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14330-1-AP-20UL
 View more versions of this product
Product Code. 16892914
  • £135.00 / 20µL
Description
This gene encodes the enzyme responsible for RNA editing by site-specific deamination of adenosines. This enzyme destabilizes double-stranded RNA through conversion of adenosine to inosine. Mutations in this gene have been associated with dyschromatosis symmetrica hereditaria. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
ADAR1
Polyclonal
Unconjugated
ADAR
ADAR, ADAR1, adenosine deaminase, RNA, DRADA, DSH, DSRAD, G1P1, IFI 4, IFI4, Interferon inducible protein 4, K88dsRBP, p136
Rabbit
Antigen Affinity Chromatography
RUO
100523784, 103, 56417, 81635
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P55265, P55266, Q99MU3
Adar
ADAR1 Fusion Protein Ag5609
20 μL
Primary
Pig, Human, Mouse, Rat