missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adar, Sheep anti-Rat, Polyclonal Antibody, Abnova™
Sheep Polyclonal Antibody
Brand: Abnova PAB9859.50ug
This item is not returnable.
View return policy
Specifications
| Adar | |
| Polyclonal | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| adenosine deaminase, RNA-specific | |
| Adar | |
| Recombinant GST fusion protein corresponding to amino acids 1116-1176 of rat Adar. | |
| RUO | |
| 81635 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| In PBS (0.08% sodium azide) | |
| - | |
| Sheep | |
| 50 μg | |
| Primary | |
| Rat | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction