Learn More
ADAR Antibody (CL0176) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05500AF405
Product Code. 30296195
  • £558.00 / 0.10mL
Description
ADAR Monoclonal antibody specifically detects ADAR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ADAR
Monoclonal
Alexa Fluor 405
ADAR1DSH, adenosine deaminase, RNA-specific, DRADAP136, DSRADadenosine deaminase acting on RNA 1-A, EC 3.5.4, EC 3.5.4.-, G1P1double-stranded RNA-specific adenosine deaminase, IFI-4, IFI4dsRNA adenosine deaminase, interferon-induced protein 4, Interferon-inducible protein 4, K88DSRBP, p136,136 kDa double-stranded RNA-binding protein
This antibody was developed using a recombinant protein derived from P55265, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL0176
50mM Sodium Borate
Mouse
Protein A purified
RUO
103
Store at 4⪚C in the dark.
IgG1