missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £387.00
Specifications
| Antigen | ADAR |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699991
|
Novus Biologicals
NBP2-92918-0.02ml |
0.02 mL |
£171.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642782
|
Novus Biologicals
NBP2-92918-0.1ml |
0.1 mL |
£387.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAR Polyclonal antibody specifically detects ADAR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| ADAR | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ADAR1DSH, adenosine deaminase, RNA-specific, DRADAP136, DSRADadenosine deaminase acting on RNA 1-A, EC 3.5.4, EC 3.5.4.-, G1P1double-stranded RNA-specific adenosine deaminase, IFI-4, IFI4dsRNA adenosine deaminase, interferon-induced protein 4, Interferon-inducible protein 4, K88DSRBP, p136,136 kDa double-stranded RNA-binding protein | |
| A synthetic peptide corresponding to a sequence within amino acids 200-299 of human ADAR1 (NP_001102.2). RPDGHSQGAPNSDPSLE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 103 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title