Learn More
Trajan Scientific and Medical™ Adaptors for 1/16 in. Female Swagelok™ Fittings
Gives low dead volume connections for optimum chromatography.
Brand: Trajan Scientific and Medical™ 103461
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Leak free connections
- Replacement parts such as nuts and ferrules
Specifications
1/16 in. Adaptor for Female Swagelok Fittings | |
Stainless Steel |