Learn More
BOLA Adaptor Tri-Clamp Hose Connector
Product Code. 16606936
Click to view available options
Diameter (Metric) Outer:
10.4 mm
Unit Size:
Each
Product Code. 16606936
Brand: BOLA D91502
Only null left
Add to basket
Add to basket
Adaptor Tri-Clamp Hose Connector Fitting for transition of Tri-Clamp as per DIN 32 676 (Plate 25,0 mm) to Hose Connector O.D. 10,4 mm
Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a hose connector. Universal chemical resistance, the flowing product is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Diameter (Metric) Outer 10.4 mm
Color White