missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Adapters for HIGHConic III Rotor

Assorted adapters to be used with the HIGHConic III rotor

Brand:  Thermo Scientific™ 75005804

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15738578

  • £144.00 / Pack of 2
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Accommodate conical tubes in the 50mL buckets for use with the Thermo Scientific HIGHConic III fixed angle rotor.

  • Adapters to run various tubes in HIGHConic III rotor
Specifications

Specifications

Rotor Adapter
Polypropylene
16 mm
HIGHConic III rotor (75005709)
1
2
Round Bottom
12 mL
96 mm
Adapters for 12 mL round bottom tubes used in HIGHConic III rotor
12 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers