missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Adapters for Fisherbrand™ accuSpin™ 4-Place Rotor with Rectangular Bucket

Product Code. 10044900 Shop All Thermo Scientific Products
Change view
Click to view available options
Container Type:
Bottle
Color:
Brown
Gray
White
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type Color unitSize
10044900 Bottle Brown Each
10612422 Bottle White Each
10371092 Bottle Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10044900 Supplier Fisherbrand™ Supplier No. 75005387

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Adapters are sold individually and are available in other sizes for convience.

Fit in rectangular bucket 13-100-587 for swinging bucket rotor 13-100-584; also can be used in sealed rotor 13-100-589.

Order Info

Adapters are sold individually. Order correct quantity to outfit rotor. Other adapter sizes are available online.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) accuSpin 1/1R 4-Place Rotor
Color Brown
Container Type Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.