Learn More
Lenz Laborglasintrumente Adapters, Bent, for Distilling Receivers Acc. to Bredt
Product Code. 10569971
Click to view available options
Standard Taper Joint:
Upper Socket NS 14/23, Lower Socket NS 19/26
Unit Size:
Each
Product Code. 10569971
Brand: Lenz Laborglasintrumente 05285101
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Adapters, bent, for distilling receivers
  • Acc. to Bredt Bent adapters (105°)
  • With two NS sockets
  • Vacuum connection: GL 18
  • Made of DURAN™ tubing
Specifications
Standard Taper Joint Upper Socket NS 14/23, Lower Socket NS 19/26