Learn More
Upchurch Scientific Adapter threaded 1/4 inch-28 flat bottom to 10-32coned PEEK(TM) 6.7µL
Brand:  Upchurch Scientific 02014-11
Adapter threaded 1/4 inch-28 flat bottom to 10-32coned PEEK(TM) 6.7µL
Product Code. 12967749
  • £22.65 / Pack of 1