Learn More
Upchurch Scientific Adapter thread to barbed 1/4 inch-24 flat bottomto 1/16 inch (1.55mm) ID ETFE natural natural
Brand:  Upchurch Scientific 02014-08
Adapter thread to barbed 1/4 inch-24 flat bottomto 1/16 inch (1.55mm) ID ETFE natural natural
Product Code. 12907749
  • £5.95 / Pack of 1