Learn More
BUCHI Glass Chromatography Column Adapter Set
Product Code. 12749505 Shop All Buchi Products
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
Product Code. Packaging Quantity unitSize
12749505 1 Ea. Each
1 options
Product Code. 12749505
Brand: BUCHI 044059
This item requires a price update prior to being ordered.
Request a Price for this item here.
Adapter for Glass Column Dry filling set for ID 70 to 100 mm
  • Dry fill silica gel into glass chromatography cartridges
  • Use medium-pressure dry gas/air to efficiently pack glass columns
  • Compatible with BUCHI glass chromatography columns
  • Alternative slurry (wet) packing columns available
Specifications
Type Cylinder
For Use With (Application) 70/100 mm Large Inlet Thread Glass Columns
Packaging Quantity 1 Ea.
Description Filling Adapter Set
Material Borosilicate Glass
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Politique de confidentialité.