Learn More
Norell™ Adapter Optimizer Insert Varian for 5mm spinner turbine 3mm Norell
Product Code. 12822326
Product Code. 12822326 Supplier Norell™ Supplier No. 5X3INSV
Only null left
Add to basket
Add to basket
Precision adapter holds 3mm, 4mm or 4.25mm NMR tube in 5mm spinner turbine.
Available for Bruker and Varian spectrometers. Does not include spinner turbine.